Biotin pentafluorophenyl ester (PFP-Biotin) ≥95%

Supplier: Ambeed
Warning

A276053-100MG A276053-250MG
103813-292EA 429 USD
103813-292 103853-222 77885-734 77885-735
Biotin pentafluorophenyl ester (PFP-Biotin) ≥95%
Biotin pentafluorophenyl ester

Precaución: Keep in a dark place, inert atmosphere, store in freezer, under -20 °C

Formula: C₁₆H₁₅F₅N₂O₃S
Boiling Pt: 555.3±50.0 °C (760 mm Hg)
Flash Pt: 289.6 °C
Storage Temperature: Freezer
MDL Number: MFCD09952636
CAS Number: 120550-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR