Pentafluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A268671-1G A268671-25G A268671-10G A268671-5G A268671-100G
103812-928EA 18 USD
103812-928 103852-830 77092-588 77092-590 77153-598 77305-012 77900-932
Pentafluorophenylboronic acid 97%
Pentafluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₂BF₅O₂
Boiling Pt: 244 °C (760 mm Hg)
Melting Pt: 176…179 °C
Flash Pt: 101.4 °C
Storage Temperature: Freezer
MDL Number: MFCD01074663
CAS Number: 1582-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR