Pentafluorophenyl 4-nitrobenzenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Pentafluorophenyl 4-Nitrobenzenesulphonate

A138784-1G A138784-5G
103802-668EA 52 USD
103802-668 77149-962 77299-022 77299-024
Pentafluorophenyl 4-nitrobenzenesulfonate 98%
Pentafluorophenyl 4-nitrobenzenesulphonate

Precaución: for Research Use Only

Formula: C₁₂H₄F₅NO₅S
MW: 369.2209 g/mol
Melting Pt: 109…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05975120
CAS Number: 244633-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR