Pentaethylene glycol monomethyl ether 95%

Supplier: Ambeed
Warning

Synonyms: mPEG5-Alcohol

A558760-10G A558760-1G A558760-5G A558760-100G
103828-740EA 8 USD
103828-740 103839-870 76726-506 76843-972 76843-974 77602-376
Pentaethylene glycol monomethyl ether 95%
Pentaethylene glycol monomethyl ether

Precaución: for Research Use Only

Formula: C₁₁H₂₄O₆
MW: 252.31 g/mol
Boiling Pt: 332.3 °C (760 mm Hg)
Flash Pt: 154.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00045995
CAS Number: 23778-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR