Pentaethylene glycol 97%

Supplier: Ambeed
Warning

Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol

A261094-100G A261094-10G A261094-500G A261094-25G A261094-5G A261094-1G
103812-520EA 9 USD
103812-520 103852-408 76722-804 76722-806 76722-808 77092-004 77092-006 77304-810
Pentaethylene glycol 97%
Pentaethylene glycol

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₆
MW: 238.28 g/mol
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002878
CAS Number: 4792-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR