4-Penten-1-ol 98%

Supplier: Ambeed
Danger

Synonyms: Pent-4-en-1-ol, 1-Penten-5-ol, 4-Penten-1-ol

A166788-10G A166788-1G A166788-25G A166788-5G A166788-100G
103806-096EA 9 USD
103806-096 103844-358 77286-630 77320-636 77320-638 77320-640 77748-101
4-Penten-1-ol 98%
4-Penten-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 141 °C (760 mm Hg)
Flash Pt: 43.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002975
CAS Number: 821-09-0
UN: 1987
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR