Pempidine 98%

Supplier: Ambeed
Danger

Synonyms: 1,2,2,6,6-Pentamethylpiperidine

A251224-1G A251224-5G A251224-100MG A251224-10G A251224-25G
103811-974EA 30 USD
103811-974 103851-816 77091-192 77091-194 77193-642 77193-644 77193-646
Pempidine 98%
Pempidine

Precaución: for Research Use Only

Formula: C₁₀H₂₁N
MW: 155.28 g/mol
Boiling Pt: 187…188 °C (760 mm Hg)
Melting Pt: 11
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00006493
CAS Number: 79-55-0
UN: 1992
ADR: 3,6.1 Ⅲ
Merck Index: 12,07207

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR