Pectolinarigenin 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5,7-Dihydroxy-6-methoxy-2-(4-methoxyphenyl)-4H-chromen-4-one
A557473-10MG
A557473-1MG
A557473-5MG
103792-768EA
142
USD
103792-768
103799-682
76991-736
76991-738
76991-740
Pectolinarigenin 95%
Pectolinarigenin
Precaución: for Research Use Only
Formula:
C₁₇H₁₄O₆ MW: 314.28 g/mol Boiling Pt: 565.5 °C (760 mm Hg) Flash Pt: 212.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00017444 CAS Number: 520-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...