N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine 99+%

Supplier: Ambeed
Warning

Synonyms: Tyrphostin AG 1517

A100205-100MG A100205-10MG A100205-1MG A100205-250MG A100205-50MG A100205-5MG
103795-294EA 56 USD
103795-294 103821-006 77025-102 77025-104 77025-106 77025-108 77025-110 77025-112
N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine 99+%
N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₄BrN₃O₂
MW: 360.2 g/mol
Boiling Pt: 472.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00934896
CAS Number: 153436-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR