XPhos Pd 98%

Supplier: Ambeed
Warning

Synonyms: Chloro(2-dicyclohexylphosphino-2′,4′,6′-triisopropyl-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II), [2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC]chloro[dicyclohexyl[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine]palladium

A148487-250MG A148487-1G A148487-25G A148487-5G A148487-10G A148487-100MG
103803-934EA 24 USD
103803-934 103842-046 76871-518 77317-504 77317-506 77317-508 77549-834 77671-086
XPhos Pd 98%
XPhos Pd

Precaución: for Research Use Only

Formula: C₄₅H₅₉ClNPPd
MW: 786.80 g/mol
Melting Pt: 202…210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20264878
CAS Number: 1310584-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR