Palladium(II) trifluoroacetate 98%

Supplier: Ambeed
Warning

Synonyms: Palladium(II) 2,2,2-trifluoroacetate

A135218-100MG A135218-10G A135218-250MG A135218-1G A135218-5G
103802-114EA 35 USD
103802-114 76719-130 76719-132 76719-134 77314-910 77314-912
Palladium(II) trifluoroacetate 98%
Palladium(II) trifluoroacetate

Precaución: for Research Use Only

Formula: C₄F₆O₄Pd
MW: 332.45 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00013204
CAS Number: 42196-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR