Ozagrel hydrochloride hydrate 97%

Supplier: Ambeed
Warning

Synonyms: Ozagrel HCl, (E)-3-(4-((1H-Imidazol-1-yl)methyl)phenyl)acrylic acid hydrochloride, trans-4-[(1-Imidazolyl)methyl]cinnamic Acid Hydrochloride, (E)-3-[4-(Imidazol-1-ylmethyl)phenyl]propenoic Acid Hydrochloride

A138148-10G A138148-25G A138148-50MG A138148-5G A138148-1G
103802-568EA 10 USD
103802-568 77137-204 77137-206 77137-208 77315-590 77685-186
Ozagrel hydrochloride hydrate 97%
Ozagrel hydrochloride hydrate

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₂
MW: 264.71 g/mol
Boiling Pt: 468 °C (760 mm Hg)
Flash Pt: 236.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06795644
CAS Number: 78712-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR