Diethylene glycol bis(p-toluenesulfonate) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Oxybis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), Diethylene Glycol Ditosylate, Bis(2-tosyloxyethyl) Ether, Bis[2-(p-toluenesulphonyloxy)ethyl] Ether
A254688-1G
A254688-5G
A254688-100G
A254688-10G
A254688-25G
A254688-500G
103812-152EA
5
USD
103812-152
103852-006
77091-444
77091-446
77289-110
77289-112
77289-114
77289-116
Diethylene glycol bis(p-toluenesulfonate) 97%
Diethylene glycol bis(p-toluenesulfonate)
Precaución: for Research Use Only
Formula:
C₁₈H₂₂O₇S₂ MW: 414.49 g/mol Melting Pt: 87…89 °C Flash Pt: 305.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026001 CAS Number: 7460-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...