Oxetane-3-carboxylic acid 95%

Supplier: Ambeed
Danger

A300021-5G A300021-250MG A300021-100MG
103814-254EA 55 USD
103814-254 103835-404 76838-096 77289-956 77571-836
Oxetane-3-carboxylic acid 95%
Oxetane-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₆O₃
Boiling Pt: 244.2 °C (760 mm Hg)
Flash Pt: 115.1 °C
Storage Temperature: Freezer
MDL Number: MFCD08669505
CAS Number: 114012-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR