Oxetan-3-ylmethanol 96%

Supplier: Ambeed
Danger

Synonyms: 3-(Hydroxymethyl)oxetane, 3-Oxetanemethanol, Oxetan-3-ylmethanol

A155942-100MG A155942-1G A155942-250MG
103804-814EA 29 USD
103804-814 103843-016 76973-310 77318-760 77611-486
Oxetan-3-ylmethanol 96%
Oxetan-3-ylmethanol

Precaución: for Research Use Only

Formula: C₄H₈O₂
MW: 88.11
Boiling Pt: 204.4 °C (760 mm Hg)
Flash Pt: 106.6 °C
Storage Temperature: Freezer
MDL Number: MFCD17015894
CAS Number: 6246-06-6
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR