oxetan-3-ylmethanamine 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Aminomethyl)oxetane, Oxetan-3-ylmethanamine

A220304-100MG A220304-250MG A220304-1G A220304-5G
103818-848EA 99 USD
103818-848 103849-730 77030-784 77030-786 77303-546 77303-548
oxetan-3-ylmethanamine 98%
oxetan-3-ylmethanamine

Precaución: for Research Use Only

Formula: C₄H₉NO
Boiling Pt: 131 °C (760 mm Hg)
Flash Pt: 62.7 °C
Storage Temperature: Freezer
MDL Number: MFCD12547197
CAS Number: 6246-05-5
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR