Oxetan-3-yl 4-methylbenzenesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: p-Toluenesulphonic acid 3-Oxetanyl ester, 3-Oxetanyl p-Toluenesulphonate, 3-Oxetanyl Tosylate

A112783-100G A112783-10G A112783-25G A112783-1G A112783-5G
103798-106EA 9 USD
103798-106 103823-942 77094-088 77094-090 77094-092 77310-766 77310-768
Oxetan-3-yl 4-methylbenzenesulfonate 97%
Oxetan-3-yl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄S
MW: 228.27 g/mol
Boiling Pt: 371.6±35.0 °C (760 mm Hg)
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD08544401
CAS Number: 26272-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR