Oxetan-3-ol 96%

Supplier: Ambeed
Danger

Synonyms: 3-Hydroxyoxetane

A110967-100G A110967-10G A110967-1G
103797-764EA 7 USD
103797-764 103823-572 77107-934 77107-936 77284-074
Oxetan-3-ol 96%
Oxetan-3-ol

Precaución: for Research Use Only

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 153.124 °C (760 mm Hg)
Flash Pt: 77.266 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09056790
CAS Number: 7748-36-9
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR