2-Hydroxymethyloxetane 95% (by GC)

Supplier: Ambeed
Warning

Synonyms: 2-Oxetanemethanol, Oxetan-2-ylmethanol, 2,4-Epoxybutanol

A116277-100MG A116277-250MG A116277-1G
103798-704EA 45 USD
103798-704 103824-566 76971-198 76971-200 77311-382
2-Hydroxymethyloxetane 95% (by GC)
2-Hydroxymethyloxetane

Precaución: for Research Use Only

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 168.8 °C (760 mm Hg)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142933
CAS Number: 61266-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR