Oxazolo[5,4-b]pyridine 97%

Supplier: Ambeed
Warning

A107891-100MG A107891-250MG A107891-1G
103797-258EA 226 USD
103797-258 76982-524 76982-526 77309-876
Oxazolo[5,4-b]pyridine 97%
Oxazolo[5,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD13175184
CAS Number: 273-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR