Oxazolo[4,5-c]pyridin-2-amine 98%

Supplier: Ambeed
Warning

A101802-100MG A101802-250MG A101802-1G
103796-144EA 206 USD
103796-144 103821-832 76821-258 76821-260 77308-730
Oxazolo[4,5-c]pyridin-2-amine 98%
Oxazolo[4,5-c]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₅N₃O
MW: 135.12 g/mol
Boiling Pt: 324 °C (760 mm Hg)
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD15526655
CAS Number: 114498-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR