Oxazolidine-2,4-dione 97%

Supplier: Ambeed
Warning

A104567-100MG A104567-250MG
103796-734EA 91 USD
103796-734 77245-320 77245-322
Oxazolidine-2,4-dione 97%
Oxazolidine-2,4-dione

Precaución: for Research Use Only

Formula: C₃H₃NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD02691902
CAS Number: 2346-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR