4-Oxazolecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Oxazole-4-carboxylic acid
A239563-1G
A239563-25G
A239563-5G
A239563-100G
A239563-250MG
103811-296EA
19
USD
103811-296
103851-094
77090-230
77090-232
77090-234
77165-736
77165-738
4-Oxazolecarboxylic acid 97%
4-Oxazolecarboxylic acid
Precaución: for Research Use Only
Formula:
C₄H₃NO₃ MW: 113.0715 g/mol Melting Pt: 142 °C Flash Pt: 128.779 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06797151 CAS Number: 23012-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...