Oxazol-5-ylmethanamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Aminomethyl)-1,3-oxazole monohydrochloride

A233285-250MG A233285-100MG A233285-5G
103810-878EA 169 USD
103810-878 77165-596 77892-393 77892-394
Oxazol-5-ylmethanamine hydrochloride 98%
Oxazol-5-ylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06804578
CAS Number: 1196156-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR