2-Aminooxazole 97%

Supplier: Ambeed
Warning

Synonyms: Oxazol-2-amine

A153199-10G A153199-250MG A153199-1G A153199-25G A153199-5G
103804-472EA 13 USD
103804-472 103842-642 77095-866 77095-868 77318-276 77318-278 77318-280
2-Aminooxazole 97%
2-Aminooxazole

Precaución: for Research Use Only

Formula: C₃H₄N₂O
MW: 84.10 g/mol
Boiling Pt: 186.6 °C (760 mm Hg)
Melting Pt: 90…95 °C
Flash Pt: 66.702 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07364485
CAS Number: 4570-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR