Oxaliplatin 98%

Supplier: Ambeed
Warning

Synonyms: (trans-1,2-Cyclohexanediamine)oxalatoplatinum(II), Oxalato(trans-l-1,2-cyclohexanediamine)platinum(II)

A100048-1MG A100048-250MG A100048-1G
103795-222EA 79 USD
103795-222 103799-192 103820-944 76717-466 76717-468 77296-554
Oxaliplatin 98%
Oxaliplatin

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O₄Pt
MW: 397.29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866327
CAS Number: 61825-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR