O-Tritylhydroxylamine 97%

Supplier: Ambeed
Warning

A136056-100G A136056-10G A136056-250MG A136056-1G A136056-25G A136056-5G
103802-268EA 6 USD
103802-268 76719-194 76719-196 76719-198 77315-156 77315-158 77315-160
O-Tritylhydroxylamine 97%
O-Tritylhydroxylamine

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO
Boiling Pt: 443.2 °C (760 mm Hg)
Melting Pt: 79…81 °C
Flash Pt: 235.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042818
CAS Number: 31938-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR