Ornidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Chloro-2-hydroxypropyl)-2-methyl-5-nitroimidazole, 1-Chloro-3-(2-methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol

A213162-25G A213162-5G A213162-500G
103848-972EA 5 USD
103848-972 77087-456 77303-274 77667-786
Ornidazole 98%
Ornidazole

Precaución: for Research Use Only

Formula: C₇H₁₀ClN₃O₃
MW: 219.63 g/mol
Boiling Pt: 443.2 °C (760 mm Hg)
Melting Pt: 85…90 °C
Flash Pt: 221.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00057960
CAS Number: 16773-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR