3,5-Dihydroxytoluene 98%

Supplier: Ambeed
Warning

Synonyms: Orcinol, 5-Methylbenzene-1,3-diol, 5-Methylresorcinol

A412104-25G A412104-5G A412104-100G A412104-10G A412104-500G
103838-286EA 22 USD
103838-286 76841-826 76841-828 76979-162 76979-164 77613-256
3,5-Dihydroxytoluene 98%
3,5-Dihydroxytoluene

Precaución: For research use only

Formula: C₇H₈O₂
MW: 124.14 g/mol
Melting Pt: 106…112 °C
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00002291
CAS Number: 504-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR