O-Phenylhydroxylamine hydrochloride 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Hydroxylamine, O-phenyl-, hydrochloride, Phenoxyamine hydrochloride, N-phenoxyamine hydrochloride
A229653-250MG
A229653-1G
A229653-5G
A229653-10G
103819-486EA
30
USD
103819-486
103850-388
76722-220
77089-236
77089-238
77593-484
O-Phenylhydroxylamine hydrochloride 97%
O-Phenylhydroxylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₈ClNO MW: 145.586 g/mol Melting Pt: 132 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043271 CAS Number: 6092-80-4 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...