Omeprazole sodium 98+%
Supplier: Ambeed
|
Danger
|
A208846-10G
A208846-25G
A208846-1G
A208846-100G
103817-890EA
10
USD
103817-890
103848-578
76721-644
77086-908
77303-076
77879-299
Omeprazole sodium 98+%
Omeprazole sodium
Precaución: for Research Use Only
Formula:
C₁₇H₁₈N₃NaO₃S Melting Pt: 220 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07802816 CAS Number: 95510-70-6 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...