Omeprazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
A449839-100G
A449839-10G
A449839-5G
103838-852EA
6
USD
103838-852
77143-514
77143-516
77307-356
Omeprazole 98%
Omeprazole
Precaución: for Research Use Only
Formula:
C₁₇H₁₉N₃O₃S MW: 345.42 g/mol Boiling Pt: 600 °C (760 mm Hg) Melting Pt: 156 °C Flash Pt: 316.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00083192 CAS Number: 73590-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...