Olsalazine Sodium 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Mordant Yellow 5, Disodium 5,5'-(Diazene-1,2-diyl)bis(2-hydroxybenzoate), 5,5'-Azobis(2-hydroxybenzoic acid sodium salt), 4,4'-Dihydroxyazobenzene-3,3'-dicarboxylic acid disodium salt
A236433-1MG
A236433-25MG
A236433-50MG
A236433-5G
A236433-1G
103811-090EA
28
USD
103811-090
103850-862
76722-338
76722-340
76722-342
76722-344
76722-346
77089-906
77304-046
77304-048
Olsalazine Sodium 97%
Olsalazine Sodium
Precaución: for Research Use Only
Formula:
C₁₄H₈N₂Na₂O₆ Boiling Pt: 557.9 °C (760 mm Hg) Melting Pt: 240 °C Flash Pt: 291.2 °C Storage Temperature: Freezer |
MDL Number:
MFCD00013300 CAS Number: 6054-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...