2-(Ammoniooxy)propane chloride 98%

Supplier: Ambeed
Warning

Synonyms: O-propan-2-ylhydroxylamine hydrochloride, O-Isopropylhydroxylamine hydrochloride

A276742-100G A276742-10G A276742-1G A276742-25G A276742-5G A276742-250MG
103813-322EA 15 USD
103813-322 103853-258 77002-076 77002-078 77093-118 77093-120 77093-122 77672-422
2-(Ammoniooxy)propane chloride 98%
2-(Ammoniooxy)propane chloride

Precaución: for Research Use Only

Formula: C₃H₁₀ClNO
MW: 111.5706 g/mol
Melting Pt: 94…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00231457
CAS Number: 4490-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR