O-Ethylhydroxylamine hydrochloride 98%

Supplier: Ambeed
Danger

A245975-1000G A245975-500G A245975-25G
103809-750EA 128 USD
103809-750 76801-426 76801-428 77304-374
O-Ethylhydroxylamine hydrochloride 98%
O-Ethylhydroxylamine hydrochloride

Precaución: for Research Use Only

Formula: C₂H₈ClNO
Melting Pt: 125…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012956
CAS Number: 3332-29-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR