1-Octylboronic acid 97%
Supplier: Ambeed
|
Warning
|
A200965-1G
A200965-25G
A200965-10G
103817-254EA
9
USD
103817-254
103847-890
77085-952
77085-954
77567-908
1-Octylboronic acid 97%
1-Octylboronic acid
Precaución: For research use only.
Formula:
C₈H₁₉BO₂ Boiling Pt: 262.6 °C (760 mm Hg) Melting Pt: 81…85 °C Flash Pt: 112.6 °C Storage Temperature: Freezer |
MDL Number:
MFCD01074560 CAS Number: 28741-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...