Octanohydroxamic acid 99%

Supplier: Ambeed
Danger

Synonyms: N-Hydroxyoctanamide, Caprylohydroxamic acid

A166310-100G A166310-10G A166310-500G A166310-5G A166310-25G
103844-310EA 5 USD
103844-310 76690-054 76690-056 76690-058 77300-824 77320-562
Octanohydroxamic acid 99%
Octanohydroxamic acid

Precaución: for Research Use Only

Formula: C₈H₁₇NO₂
MW: 159.2261 g/mol
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00143918
CAS Number: 7377-03-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR