3-Azabicyclo[3.3.0]octane hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Octahydrocyclopenta[c]pyrrole hydrochloride

A234560-25G A234560-100G A234560-1G A234560-5G A234560-10G A234560-500G
103810-958EA 7 USD
103810-958 103850-714 77089-704 77113-108 77303-982 77303-984 77685-647 77685-648
3-Azabicyclo[3.3.0]octane hydrochloride 97%
3-Azabicyclo[3.3.0]octane hydrochloride

Precaución: for Research Use Only

Formula: C7H14ClN
MW: 147.65 g/mol
Boiling Pt: 204.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06411019
CAS Number: 112626-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR