Octahydro-2H-chromen-2-one 97%

Supplier: Ambeed
Warning

A336683-5G A336683-100G A336683-10G A336683-1G
103836-704EA 21 USD
103836-704 76839-710 77251-336 77251-338 77251-340
Octahydro-2H-chromen-2-one 97%
Octahydro-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₉H₁₄O₂
Boiling Pt: 145 °C (16 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00221714
CAS Number: 4430-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR