2-Bromobenzaldehyde 98%

Supplier: Ambeed
Warning

A771243-25G A771243-1000G A771243-100G A771243-10G A771243-500G
103840-934EA 6 USD
103840-934 76845-182 77118-250 77118-252 77118-254 77118-256
2-Bromobenzaldehyde 98%
2-Bromobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrO
Boiling Pt: 230.8 °C (760 mm Hg)
Melting Pt: 21…22 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00003300
CAS Number: 6630-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR