HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 98%

Supplier: Ambeed
Warning

Synonyms: HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)

A833302-100G A833302-500G A833302-25G A833302-5G
103810-582EA 85 USD
103810-582 103841-178 76880-460 76880-462 77308-422 77308-424
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 98%
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)

Precaución: for Research Use Only

Formula: C₁₁H₁₆F₆N₅OP
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075445
CAS Number: 94790-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR