O-Allylhydroxylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: O-Allylhydroxylamine Hydrochloride

A197397-100G A197397-10G A197397-1G A197397-25G A197397-5G
103816-916EA 11 USD
103816-916 103847-538 76721-344 76721-346 77085-542 77085-544 77085-546
O-Allylhydroxylamine hydrochloride 97%
O-Allylhydroxylamine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₇NO
MW: 109.55 g/mol
Boiling Pt: 123.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00012957
CAS Number: 38945-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR