O-(Tetrahydropyran-2-yl)-hydroxylamine 98%

Supplier: Ambeed
Warning

A193890-10G A193890-100G
103816-596EA 9 USD
103816-596 103847-204 77567-808 77885-439
O-(Tetrahydropyran-2-yl)-hydroxylamine 98%
O-(Tetrahydropyran-2-yl)-hydroxylamine

Precaución: For research use only. Keep in a dark place, inert atmosphere, store in freezer, under −20 °C

Formula: C₅H₁₁NO₂
Boiling Pt: 209.3 °C (760 mm Hg)
Melting Pt: 34…37 °C
Flash Pt: 82 °C
Storage Temperature: Freezer
MDL Number: MFCD01321374
CAS Number: 6723-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR