O-(4-Nitrobenzoyl)hydroxylamine 98%
Supplier: Ambeed
|
Warning
|
A307666-100G
A307666-10G
A307666-250MG
A307666-500G
A307666-1G
A307666-25G
A307666-5G
103814-514EA
6
USD
103814-514
103835-692
76816-846
76816-848
76816-850
76816-852
76838-436
76838-438
76838-440
O-(4-Nitrobenzoyl)hydroxylamine 98%
O-(4-Nitrobenzoyl)hydroxylamine
Precaución: for Research Use Only
Formula:
C₇H₆N₂O₄ MW: 182.14 g/mol Melting Pt: 108…111 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11976095 CAS Number: 35657-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...