1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride 97%

Supplier: Ambeed
Danger

A148293-100G A148293-100MG A148293-10G A148293-250MG A148293-1G A148293-25G A148293-5G
103803-908EA 9 USD
103803-908 103842-024 77190-680 77190-682 77190-684 77190-686 77317-468 77317-470 77317-472
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride 97%
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNO₂
Boiling Pt: 286.5 °C (760 mm Hg)
Melting Pt: 209…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01114582
CAS Number: 876-33-5
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR