1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride 97%

Supplier: Ambeed
Danger

A213304-100G A213304-1G A213304-25G A213304-5G
103818-288EA 11 USD
103818-288 103848-994 76999-668 77087-500 77087-502 77087-504
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride 97%
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride

Precaución: for Research Use Only

Formula: C₇H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD01114581
CAS Number: 51572-89-5
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR