1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride 97%

Supplier: Ambeed
Danger

Synonyms: O-(4-(Trifluoromethyl)benzyl)hydroxylamine hydrochloride

A150714-100MG A150714-250MG A150714-1G
103804-192EA 15 USD
103804-192 76997-326 76997-328 77299-820
1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride 97%
1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride

Precaución: for Research Use Only

Formula: C₈H₉ClF₃NO
MW: 227.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00231881
CAS Number: 321574-29-2
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR