tert-Butyl-N-hydroxycarbamate 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine

A197611-100G A197611-500G A197611-25G
103809-326EA 155 USD
103809-326 103847-574 76986-338 76986-340 77302-560
tert-Butyl-N-hydroxycarbamate 98%
tert-Butyl-N-hydroxycarbamate

Precaución: for Research Use Only

Formula: C₅H₁₁NO₃
MW: 133.15 g/mol
Boiling Pt: 250.4 °C (760 mm Hg)
Melting Pt: 53…55 °C
Flash Pt: 105.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00002107
CAS Number: 36016-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR