BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester) 98%

Supplier: Ambeed
Warning

Synonyms: N-(3-Maleimidopropionyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate, N-Succinimidyl 3-Maleimidopropionat, 3-Maleimidopropionic Acid N-Succinimidyl Ester, N-Succinimidyl 3-maleimidopropionate, 3-(Maleimido)propionic acid N-hydroxysuccinimide ester, N-β-maleimidopropyl-oxysuccinimide ester, BMPS

A395905-250MG A395905-1G A395905-5G A395905-10G
103826-972EA 8 USD
103826-972 103838-010 76725-042 76841-432 76841-434 77551-186
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester) 98%
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₆
MW: 266.21 g/mol
Boiling Pt: 448.8 °C (760 mm Hg)
Melting Pt: 168…170 °C
Flash Pt: 225.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043141
CAS Number: 55750-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR