2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Prop-2-ynyl)isoindoline-1,3-dione, N-Propargylphthalimide, N-(2-Propynyl)phthalimide

A465845-1G A465845-5G A465845-25G
103827-976EA 7 USD
103827-976 103839-050 76842-958 76842-960 77307-446
2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione 98%
2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione

Precaución: for Research Use Only

Formula: C₁₁H₇NO₂
MW: 185.18 g/mol
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00065028
CAS Number: 7223-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR